Learn More
Saint Gobain Life Sciences Chemware™ PTFE Forceps
Chemware™ PTFE Forceps for laboratory work involving hot, cold, delicate, or highly reactive materials.
Brand:  Saint Gobain Life Sciences D1069095
Product Code. 12351369
  • £382.00 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from PTFE resin with an encapsulated metal insert for strength
  • Chemically inert and noncontaminating
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5.87 in.
149 mm
1 Ea.
PTFE
Forceps