Learn More
Thermo Scientific Chemicals Salbutamol hemisulfate, 98%
£75.27 - £208.04
Chemical Identifiers
CAS | 51022-70-9 |
---|---|
Molecular Formula | C13H21NO3·1/2H2SO4 |
Molecular Weight (g/mol) | 288.35 |
InChI Key | OVICLFZZVQVVFT-UHFFFAOYSA-N |
Synonym | salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate |
PubChem CID | 9884233 |
IUPAC Name | 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid |
SMILES | CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15443913
|
Thermo Scientific Chemicals
456460010 |
1 g |
£75.27
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15565196
|
Thermo Scientific Chemicals
456460050 |
5 g |
£208.04
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51022-70-9 | |
288.35 | |
salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate | |
4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid |
C13H21NO3·1/2H2SO4 | |
OVICLFZZVQVVFT-UHFFFAOYSA-N | |
9884233 | |
CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O |
Specifications
51022-70-9 | |
Authentic | |
Glass Bottle | |
salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate | |
OVICLFZZVQVVFT-UHFFFAOYSA-N | |
4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid | |
9884233 | |
98% |
180.0°C | |
97.5% min. (HPLC) | |
C13H21NO3·1/2H2SO4 | |
Solubility in water: soluble. | |
CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O | |
288.35 | |
288.35 | |
Salbutamol hemisulfate |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 256-916-8
- Salbutamol hemisulfate
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H351-Suspected of causing cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only