missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SalI (10 U/μL)

Product Code. p-4522050 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1500 U
2,000 units
5 x 1,500 units
Unit Size:
1500 units
2000 units
Pack of 5
This item is not returnable. View return policy

Product Code. 10242440

Brand: Thermo Scientific™ ER0642

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The SalI restriction enzyme recognizes G^TCGAC sites and cuts best at 37°C in O buffer.

Lambda DNA digested with SalI, 0.7% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G▵T C G A C...3'

3'...C A G C T▵G...5'

Conditions for 100% Activity

  • 1X Buffer O:50mM Tris-HCl (pH7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer

  • SalI is supplied in:10mM Tris-HCl (pH7.4 at 25°C),100mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage

  • After 50-fold overdigestion with SalI, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: completely overlaps — blocked
  • EcoKI: never overlaps — no effect
  • EcoBI: never overlaps — no effect

Digestion of Agarose-embedded DNA

  • Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours

Compatible Ends

  • Eco88I, SmoI, XhoI

Note:

Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity. Supercoiled forms of pBR322 and pUC require 10-fold overdigestion with SalI to achieve complete digestion. Incubation at 25°C results in 50-75% activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Sal I
Compatible Buffer 10X Buffer O
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 5 x 1,500 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.