missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, 99%

Product Code. 10071290 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 g
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
1kg
250g
2g
5kg
This item is not returnable. View return policy

Product Code. 10071290

Brand: Thermo Scientific Acros 132601000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-02-8 | C7H6O2 | 122.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-02-8, 108-95-2
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O

Specifications

Melting Point -7.0°C
Color Yellow
Density 1.1600g/mL
Boiling Point 197.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. (Phenol) (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CHO
Refractive Index 1.5720 to 1.5740
Quantity 100 g
Beilstein 08,31
Merck Index 15,8462
Specific Gravity 1.16
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 122.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Salicylaldehyde
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Salicylaldehyde
Signal Word
  • Danger||Warning
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P391-Collect spillage.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Salicylaldehyde, Phenol

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt