missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylaldehyde azine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
Solubility
Soluble in dimethyl sulfoxide, methanol and ethyl acetate. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
Solubility
Soluble in dimethyl sulfoxide, methanol and ethyl acetate. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 216°C to 222°C |
Assay Percent Range | 97% |
Quantity | 50 g |
Beilstein | 1913597 |
Solubility Information | Soluble in dimethyl sulfoxide,methanol and ethyl acetate. Slightly soluble in water. |
Formula Weight | 240.26 |
Percent Purity | 97% |
Chemical Name or Material | Salicylaldehyde azine |
Safety and Handling
Product Identifier
- Salicylaldehyde azine
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'