missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldoxime, 98%, Thermo Scientific Chemicals

Product Code. 11458943
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-67-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00002120
InChI Key GFCNBJDXQMZOOC-WAYWQWQTSA-N
Synonym salicylaldoxime, 2-hydroxybenzaldehyde oxime, salicylaldehyde oxime, 2-hydroxyimino methyl phenol, 6-hydroxyamino methylidene cyclohexa-2,4-dien-1-one, e-2-hydroxybenzaldehyde oxime, orihzizptztncu-uhfffaoysa-n, orihzizptztncu-vmpitwqzsa-n, 2-1e-hydroxyimino methyl phenol, salicylideneaminoalcohol
PubChem CID 6740756
IUPAC Name 6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
SMILES ON\C=C1\C=CC=CC1=O

Specifications

Melting Point 55°C to 59°C
Quantity 25 g
Beilstein 1859881
Merck Index 14,8327
Formula Weight 137.14
Percent Purity 98%
Chemical Name or Material Salicylaldoxime

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'