Learn More
Salicylaldoxime, 98%, Thermo Scientific Chemicals
£29.90 - £390.00
Chemical Identifiers
CAS | 94-67-7 |
---|---|
Molecular Formula | C7H7NO2 |
Molecular Weight (g/mol) | 137.14 |
MDL Number | MFCD00002120 |
InChI Key | GFCNBJDXQMZOOC-WAYWQWQTSA-N |
Synonym | salicylaldoxime, 2-hydroxybenzaldehyde oxime, salicylaldehyde oxime, 2-hydroxyimino methyl phenol, 6-hydroxyamino methylidene cyclohexa-2,4-dien-1-one, e-2-hydroxybenzaldehyde oxime, orihzizptztncu-uhfffaoysa-n, orihzizptztncu-vmpitwqzsa-n, 2-1e-hydroxyimino methyl phenol, salicylideneaminoalcohol |
PubChem CID | 6740756 |
IUPAC Name | 6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one |
SMILES | ON\C=C1\C=CC=CC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10062790
|
Thermo Scientific Acros
207400250 |
25 g | Glass bottle |
£29.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10216790
|
Thermo Scientific Acros
207401000 |
100 g | Glass bottle |
£92.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10056143
|
Thermo Scientific Acros
207405000 |
500 g | Glass bottle |
£390.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
94-67-7 | |
137.14 | |
GFCNBJDXQMZOOC-WAYWQWQTSA-N | |
6740756 | |
ON\C=C1\C=CC=CC1=O |
C7H7NO2 | |
MFCD00002120 | |
salicylaldoxime, 2-hydroxybenzaldehyde oxime, salicylaldehyde oxime, 2-hydroxyimino methyl phenol, 6-hydroxyamino methylidene cyclohexa-2,4-dien-1-one, e-2-hydroxybenzaldehyde oxime, orihzizptztncu-uhfffaoysa-n, orihzizptztncu-vmpitwqzsa-n, 2-1e-hydroxyimino methyl phenol, salicylideneaminoalcohol | |
6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one |
Specifications
94-67-7 | |
Bordeaux to Purple-Red | |
97.5% min. (GC) | |
2-(HO)C6H4CH=NOH | |
08, 49 | |
salicylaldoxime, 2-hydroxybenzaldehyde oxime, salicylaldehyde oxime, 2-hydroxyimino methyl phenol, 6-hydroxyamino methylidene cyclohexa-2,4-dien-1-one, e-2-hydroxybenzaldehyde oxime, orihzizptztncu-uhfffaoysa-n, orihzizptztncu-vmpitwqzsa-n, 2-1e-hydroxyimino methyl phenol, salicylideneaminoalcohol | |
ON\C=C1\C=CC=CC1=O | |
137.14 | |
137.14 | |
Powder |
56°C to 59°C | |
Authentic | |
C7H7NO2 | |
MFCD00002120 | |
15, 8464 | |
GFCNBJDXQMZOOC-WAYWQWQTSA-N | |
6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one | |
6740756 | |
98% | |
Salicylaldoxime |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-353-8
- Salicylaldoxime
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only