missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylamide, 99%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 139.0°C to 142.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (on anhydrous substance) |
| Packaging | Plastic bottle |
| Linear Formula | C6H4(OH)CONH2 |
| Quantity | 250 g |
| Beilstein | 10, 87 |
| Merck Index | 15, 8465 |
| Solubility Information | Solubility in water: soluble in hot water. Other solubilities: 0.2% in water (30°C), 10.0% in propylene glycol (5°C), soluble in alcohol, chloroform and ether, slightly soluble in carbon tetrachloride |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction