Learn More
Salicylamide, 99%, Thermo Scientific Chemicals
£18.10 - £63.70
Chemical Identifiers
CAS | 65-45-2 |
---|---|
Molecular Formula | C7H7NO2 |
Molecular Weight (g/mol) | 137.14 |
MDL Number | MFCD00007978 |
InChI Key | SKZKKFZAGNVIMN-UHFFFAOYSA-N |
Synonym | salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin |
PubChem CID | 5147 |
ChEBI | CHEBI:32114 |
IUPAC Name | 2-hydroxybenzamide |
SMILES | NC(=O)C1=CC=CC=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10236430
|
Thermo Scientific Acros
229360050 |
5 g | Glass Bottle |
£18.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10134810
|
Thermo Scientific Acros
229362500 |
250 g | Plastic bottle |
£26.30
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10388650
|
Thermo Scientific Acros
229360010 |
1 kg | Plastic bottle |
£63.70
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
65-45-2 | |
137.14 | |
SKZKKFZAGNVIMN-UHFFFAOYSA-N | |
5147 | |
2-hydroxybenzamide |
C7H7NO2 | |
MFCD00007978 | |
salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin | |
CHEBI:32114 | |
NC(=O)C1=CC=CC=C1O |
Specifications
65-45-2 | |
White | |
98.5% min. (on anhydrous substance) | |
C6H4(OH)CONH2 | |
10, 87 | |
salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin | |
SKZKKFZAGNVIMN-UHFFFAOYSA-N | |
2-hydroxybenzamide | |
5147 | |
137.14 | |
Crystalline Powder |
139.0°C to 142.0°C | |
Authentic | |
C7H7NO2 | |
MFCD00007978 | |
15, 8465 | |
Solubility in water: soluble in hot water. Other solubilities: 0.2% in water (30°C), 10.0% in propylene glycol (5°C), soluble in alcohol, chloroform and ether, slightly soluble in carbon tetrachloride | |
NC(=O)C1=CC=CC=C1O | |
137.14 | |
CHEBI:32114 | |
99% | |
Salicylamide, 99% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-609-3
missing translation for 'rtecsNumber' : VN6475000
missing translation for 'tsca' : TSCA
RUO – Research Use Only