missing translation for 'onlineSavingsMsg'
Learn More
Salicylic Acid, 99+%, Thermo Scientific Chemicals Product Code.: 10543504

Salicylic Acid, 99+%, Thermo Scientific Chemicals

Product Code. 10543504
2.5 kg, Plastic bottle
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10543504

Brand: Thermo Scientific Acros 147700025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
Synonym salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
PubChem CID 338
ChEBI CHEBI:16914
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

Melting Point 158.0°C to 161.0°C
Boiling Point 211.0°C (20.0 mmHg)
Color White
Assay Percent Range 99+%
Beilstein 10, 43
Merck Index 15, 8469
Formula Weight 138.12
Percent Purity ≥99%
Quantity 2.5 kg
Flash Point 157°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Chemical Name or Material Salicylic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458464 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number147700025,147700010
Lot NumberA0400205 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number147700025,147700010
Lot NumberA0407386 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number147700025,147700010
Lot NumberA0408025 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number147700025,147700010
Lot NumberA0418294 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number147700025,147700010
compliance-icons compliance-icons
Product Identifier
  • Salicylic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Salicylic Acid, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.