missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylic Acid, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
Specifications
Specifications
Melting Point | 158°C |
Color | White |
pH | 3.6 |
Boiling Point | 211°C |
Packaging | Plastic powder jar |
Quantity | 250 g |
Formula Weight | 138.12g/mol |
Vapor Pressure | 0.3mbar 95 |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Salicylic acid
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'