missing translation for 'onlineSavingsMsg'
Learn More

Salicylsalicylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10336590
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-94-3
Molecular Formula C14H10O5
Molecular Weight (g/mol) 258.22
MDL Number MFCD00020252
InChI Key WVYADZUPLLSGPU-UHFFFAOYSA-N
Synonym salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal
PubChem CID 5161
ChEBI CHEBI:9014
IUPAC Name 2-(2-hydroxybenzoyl)oxybenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O

Specifications

Melting Point 139.0°C to 151.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 10,II,54
Merck Index 15,8477
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in alcohol and ether, sparingly soluble in benzene, slightly soluble in acetone
Formula Weight 258.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Salicylsalicylic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Salicylsalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'