missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Salubrinal Product Code.: 15446259

Thermo Scientific Chemicals Salubrinal

Product Code. 15446259
10 mg
Click to view available options
Quantity:
5 mg
10 mg
This item is not returnable. View return policy

Product Code. 15446259

Brand: Thermo Scientific Chemicals J64192.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Salubrinal is a selective Inhibitor of eIF2α (eukaryotic translation initiation factor 2 α-subunit) dephosphorylation, inhibiting global protein translation. Salubrinal is a specific inhibitor of ER Stress induced apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405060-95-9
Molecular Formula C21H17Cl3N4OS, C21H17Cl3N4OS
Synonym eIF-2alpha Inhibitor; SAL

Specifications

Quantity 10 mg
Description Pale Brown
Content And Storage 2°C to 8°C
Formula Weight 479.81g/mol
Sensitivity Light Sensitive
Product Type Apoptosis Reagent
Form Powder
InChI Key LCOIAYJMPKXARU-UHFFFAOYNA-N
SMILES ClC(Cl)(Cl)C(NC(=S)NC1=C2N=CC=CC2=CC=C1)NC(=O)C=CC1=CC=CC=C1
IUPAC Name 3-phenyl-N-(2,2,2-trichloro-1-{[(quinolin-8-yl)carbamothioyl]amino}ethyl)prop-2-enamide
Molecular Weight (g/mol) 479.80
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Salubrinal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.