missing translation for 'onlineSavingsMsg'
Learn More

Salvequick Pilferproof Plaster Refill

Product Code. p-8010006
Click to view available options
Material:
Fabric
Metal-detectable
Plastic
Color:
Beige
Blue
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12940475

Brand: Safety First CD7

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Material Metal-detectable
Product Type Salvequick Plaster Refill
Sterility Sterile
Quantity 6 Pk.
Packaging Type Refill
For Use With (Application) Salvequick Pilferproof Plaster Dispenser, Food Industry
Color Blue
Includes Twenty-one 72 x 19 mm; Fourteen 72 x 25 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.