missing translation for 'onlineSavingsMsg'
Learn More

Samarium foil, 1.0mm (0.04in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11396199
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in control rods in nuclear reactors, as a catalyst in organic chemistry, a pyrophoric metal in lighter flints, and a core in carbon arc-lamp electrodes. Also used in alloys with cobalt to make strong permanent magnets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mylar under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Quantity 50 x 50 mm
Solubility Information Insoluble in water.
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈4.9g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Samarium foil, 1.0mm (0.04 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'