Learn More
Samarium(III) bromide hexahydrate, 99.99%, Thermo Scientific™
£41.71 - £134.00
Chemical Identifiers
CAS | 13517-12-9 |
---|---|
Molecular Formula | Br3H12O6Sm |
Molecular Weight (g/mol) | 498.162 |
MDL Number | MFCD00192430 |
InChI Key | LXJDCZVUWJNXRL-UHFFFAOYSA-K |
Synonym | acmc-20ajrk, samarium bromide hexahydrate, samarium 3+ bromide-water 1/3/6, samarium iii bromide hexahydrate, reacton™ |
PubChem CID | 57348178 |
IUPAC Name | samarium(3+);tribromide;hexahydrate |
SMILES | O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443717
|
Thermo Scientific Alfa Aesar
A18191.09 |
10 g |
£41.71
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453717
|
Thermo Scientific Alfa Aesar
A18191.18 |
50 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13517-12-9 | |
498.162 | |
LXJDCZVUWJNXRL-UHFFFAOYSA-K | |
57348178 | |
O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3] |
Br3H12O6Sm | |
MFCD00192430 | |
acmc-20ajrk, samarium bromide hexahydrate, samarium 3+ bromide-water 1/3/6, samarium iii bromide hexahydrate, reacton™Show More | |
samarium(3+);tribromide;hexahydrate |
Specifications
>640°C | |
Br3H12O6Sm | |
acmc-20ajrk, samarium bromide hexahydrate, samarium 3+ bromide-water 1/3/6, samarium iii bromide hexahydrate, reacton™ | |
O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3] | |
498.162 | |
498.15 (390.06 Anhydrous) | |
Hygroscopic |
13517-12-9 | |
MFCD00192430 | |
LXJDCZVUWJNXRL-UHFFFAOYSA-K | |
samarium(3+);tribromide;hexahydrate | |
57348178 | |
99.99% | |
Samarium(III) bromide hexahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 237-347-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.