missing translation for 'onlineSavingsMsg'
Learn More
Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11372558

Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11372558
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11372558

Brand: Thermo Scientific Alfa Aesar 014603.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13517-12-9
Molecular Formula Br3H12O6Sm
Molecular Weight (g/mol) 498.162
MDL Number MFCD00192430
InChI Key LXJDCZVUWJNXRL-UHFFFAOYSA-K
Synonym acmc-20ajrk, samarium bromide hexahydrate, samarium 3+ bromide-water 1/3/6, samarium iii bromide hexahydrate, reacton™
PubChem CID 57348178
IUPAC Name samarium(3+);tribromide;hexahydrate
SMILES O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]

Specifications

Melting Point >640°C
Physical Form Crystalline
Quantity 100 g
Formula Weight 498.15 (390.06 Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Samarium(III) bromide hexahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS20K036 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number014603.22,014603.14
Lot NumberN14K014 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number014603.22,014603.14
Lot NumberM24J008 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog Number014603.22,014603.14
Lot NumberR26B022 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014603.22,014603.14
Lot NumberQ04F025 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014603.22,014603.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.