missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)

Product Code. 11312278
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11312278

Brand: Thermo Scientific Alfa Aesar 013177.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38245-37-3 | C3O9Sm2 | 480.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38245-37-3
Molecular Formula C3O9Sm2
Molecular Weight (g/mol) 480.74
MDL Number MFCD00150444
InChI Key QCZFMLDHLOYOQJ-UHFFFAOYSA-H
IUPAC Name disamarium(3+) tricarbonate
SMILES [Sm+3].[Sm+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point >500°C (decomposition)
Physical Form Powder
Quantity 25 g
Formula Weight 480.73 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Samarium(III) carbonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.