missing translation for 'onlineSavingsMsg'
Learn More
Samarium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11413860

Samarium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11413860
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11413860

Brand: Thermo Scientific Alfa Aesar 089916.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Samarium(III) chloride is used in laboratories, preparation of samarium metals. The anhydrous chloride is used to prepare organometallic compounds of samarium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-82-7
Molecular Formula Cl3Sm
Molecular Weight (g/mol) 256.71
MDL Number MFCD00011235
InChI Key BHXBZLPMVFUQBQ-UHFFFAOYSA-K
IUPAC Name samarium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Sm+3]

Specifications

Boiling Point (decomposition)
Melting Point 678°C
Physical Form Chunks and/or Powder
Quantity 10 g
Merck Index 14,8347
Solubility Information Soluble in water.
Formula Weight 256.71
Percent Purity 99.9%
Sensitivity Hygroscopic
Density 4.46 g/mL at 16°C
Assay (REO)
Chemical Name or Material Samarium (III) chloride, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ27K055 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number089916.18,089916.09
Lot NumberZ07J006 Certificate TypeCertificate of Analysis Date03/01/2024 Catalog Number089916.18,089916.09
Lot NumberW01J041 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number089916.18,089916.09
Lot NumberA15T006 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089916.18,089916.09
Lot NumberT01G001 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089916.18,089916.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Samarium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.