Learn More
Samarium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
£56.68 - £184.92
Chemical Identifiers
CAS | 10361-82-7 |
---|---|
Molecular Formula | Cl3Sm |
Molecular Weight (g/mol) | 256.71 |
MDL Number | MFCD00011235 |
InChI Key | BHXBZLPMVFUQBQ-UHFFFAOYSA-K |
IUPAC Name | samarium(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Sm+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316288
|
Thermo Scientific Alfa Aesar
011231.14 |
25 g |
£56.68
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306288
|
Thermo Scientific Alfa Aesar
011231.22 |
100 g |
£184.92
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Samarium(III) chloride hydrate is used for the preparation of samarium metal, which finds use as magnets. It is combined with sodium chloride to get a low melting point eutectic mixture. Further, it serves as a precursor for the preparation of other samarium salts and bis(pentamethylcyclopentadienyl)alkylsamarium(III) complexes. In addition, it serves as a catalyst for hydrogenation and hydrosilylation of alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10361-82-7 | |
256.71 | |
BHXBZLPMVFUQBQ-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[Sm+3] |
Cl3Sm | |
MFCD00011235 | |
samarium(3+) trichloride |
Spezifikation
110°C (in -5H2O) | |
Crystalline Aggregates | |
MFCD00011235 | |
Soluble in water and pyridine. | |
[Cl-].[Cl-].[Cl-].[Sm+3] | |
256.71 | |
99.9% | |
Acidic | |
(REO) |
10361-82-7 | |
Cl3Sm | |
14,8347 | |
BHXBZLPMVFUQBQ-UHFFFAOYSA-K | |
samarium(3+) trichloride | |
256.71 (Anhydrous) | |
Reacton™ | |
Hygroscopic | |
Samarium(III) chloride hydrate |
Sicherheit und Handhabung
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 233-797-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only