missing translation for 'onlineSavingsMsg'
Learn More
Samarium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11346288

Samarium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11346288
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11346288

Brand: Thermo Scientific Alfa Aesar 011232.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Samarium(III) chloride is used for the preparation of samarium metal, which has a variety of uses, notably in magnets. In laboratory Samarium(III) chloride can also be used as a starting point for the preparation of other samarium salts. It is used as infrares absorbing glass, lasers, color television phosphors, magnets, microwave filters. It is used as catalyst in cracking crude petroleum and used as an alloy with cobalt for magnets. It is used as neutron capture masers and have applications in nuclear industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-82-7
Molecular Formula Cl3Sm
Molecular Weight (g/mol) 256.71
MDL Number MFCD00011235
InChI Key BHXBZLPMVFUQBQ-UHFFFAOYSA-K
IUPAC Name samarium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Sm+3]

Specifications

Melting Point 110°C (in -5H2O)
Physical Form Wet Crystalline Aggregates
Quantity 100 g
Merck Index 14,8347
Solubility Information Soluble in water.
Formula Weight 256.71 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Acidic
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Samarium(III) chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Samarium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.