Learn More
Samarium(III) fluoride, anhydrous, REacton™, 99.99% (REO)
Description
Samarium(III) fluoride is used as laboratory reagent, in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. It has specialized uses in phosphors, lasers, and thermoelectric devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1306°C |
| Physical Form | Powder |
| Quantity | 50 g |
| UN Number | UN3288 |
| Solubility Information | Insoluble in water. Moderately soluble in strong mineral acids. |
| Formula Weight | 207.35 |
| Percent Purity | 99.99% |
| Grade | Reacton |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.