missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) hydroxide hydrate, 99% (REO)

Product Code. 11338318
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11338318

Brand: Thermo Scientific Alfa Aesar 039512.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20403-06-9 | H3O3Sm | 201.38 g/mol

Samarium hydroxide (Sm(OH)3) is an important samarium compound with many potential applications in electronics, optics and catalysis. It is used as a chemical intermediate. It is used in the separation and preconcentration procedures for the determination of lead using spectrometric techniques. It has specialized uses in glass, phosphors, lasers, and thermoelectric devices and it is mainly applied in Neutron absorption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20403-06-9
Molecular Formula H3O3Sm
Molecular Weight (g/mol) 201.38
MDL Number MFCD00050108
InChI Key BCYBEIXXOVNETJ-UHFFFAOYSA-K
IUPAC Name samarium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Sm+3]

Specifications

Physical Form Lump/Powder
Quantity 5 g
UN Number UN3262
Solubility Information Soluble in water and moderately soluble in strong mineral acids.
Formula Weight 201.37 (Anhydrous)
Percent Purity 99%
Assay (REO)
Chemical Name or Material Samarium(III) hydroxide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.