missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) nitrate hydrate, REacton™, 99.99% (REO)

Codice prodotto. 11376268
Click to view available options
Quantity:
5 g
25 g
100 g
Dimensione della confezione:
100g
25g
5g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 11376268

Marca: Thermo Scientific Alfa Aesar 011224.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 13759-83-6 | H12N3O15Sm | 444.46 g/mol

Samarium(III) nitrate hydrate is used as a catalyst in von Pechmann condensation for the preparation of coumarins. Further, it finds use in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. It is also used as a laboratory reagent. In addition to this, it is used in NMR spectroscopy for spectral resolution and is used for doping purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 13759-83-6
Molecular Formula H12N3O15Sm
Molecular Weight (g/mol) 444.46
MDL Number MFCD00149857
InChI Key HDCOFJGRHQAIPE-UHFFFAOYSA-N
Synonym samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
PubChem CID 203081
SMILES O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifica

Melting Point 78°C to 79°C
Color Yellow
pH ∼3
Physical Form Crystalline Aggregates
Quantity 5 g
UN Number UN1477
Solubility Information Soluble in water and acetonitrile.
IUPAC Name samarium(3+);trinitrate;hexahydrate
Formula Weight 336.36 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 2.375 g/mL
Assay (REO)
Chemical Name or Material Samarium(III) nitrate hydrate
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato