missing translation for 'onlineSavingsMsg'
Learn More
Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm, Thermo Scientific Chemicals Product Code.: 11351438

Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm, Thermo Scientific Chemicals

Product Code. 11351438
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11351438

Brand: Thermo Scientific Alfa Aesar 010915.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. It finds use as a neutron absorber in nuclear power reactors to control the reaction. It is also used as a catalyst for the dehydration of primary alcohols to aldehydes or ketones. It forms samarium perchlorate when treated with perchloric acid, which is used as a reference material for checking the wavelength scale of a spectrophotometer in the range of 200 to 500 nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-58-1
MDL Number MFCD00011237

Specifications

Melting Point 2300°C
Physical Form Powder
Quantity 25 g
Merck Index 14,8347
Solubility Information Insoluble in water.
Formula Weight 348.7
Percent Purity 99.998%
Grade Reacton™
Refractive Index 1.97
Assay (metals basis)
Chemical Name or Material Samarium(III) oxide, REM 100ppm
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ03H033 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010915.22,010915.14,010915.06
Lot NumberF14Y019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010915.22,010915.14,010915.06
Lot NumberK18Z002 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number010915.22,010915.14,010915.06
Lot NumberQ14A005 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number010915.22,010915.14,010915.06
Lot NumberP4362 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number010915.22,010915.14,010915.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.