missing translation for 'onlineSavingsMsg'
Learn More
Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals Product Code.: 15446335

Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Product Code. 15446335
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 15446335

Brand: Thermo Scientific Alfa Aesar 040567.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13569-60-3
Molecular Formula Cl3H2O13Sm
Molecular Weight (g/mol) 466.71
MDL Number MFCD00049769
InChI Key ONSMNAINZIBBGR-UHFFFAOYSA-K
Synonym samarium 3+ ;triperchlorate, samarium iii perchlorate solution, trihyperchloric acid samarium iii salt, samarium iii perchlorate w/w aqueous solution, reagent gradeShow MoreShow Less
IUPAC Name samarium(3+) hydrate triperchlorate
SMILES O.[Sm+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Liquid
Quantity 50 g
UN Number UN3211
Formula Weight 556.80 (448.70 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Samarium(III) perchlorate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP12G036 Certificate TypeCertificate of Analysis Date27/07/2022 Catalog Number040569.NC,040567.18,040567.09
Lot NumberP12G026 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040569.NC,040567.18,040567.09
Lot NumberE11Y031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040569.NC,040567.18,040567.09
Lot NumberL01P07 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040569.NC,040567.18,040567.09
Lot NumberE27U018 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040569.NC,040567.18,040567.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Samarium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.