missing translation for 'onlineSavingsMsg'
Learn More
Samarium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals Product Code.: 10370524

Samarium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Product Code. 10370524
10 g, Plastic bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10370524

Brand: Thermo Scientific Acros 383170100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52093-28-4
Molecular Formula C3F9O9S3Sm
Molecular Weight (g/mol) 597.55
MDL Number MFCD00192431
InChI Key DDCWGUIPLGMBPO-UHFFFAOYSA-K
Synonym samarium iii trifluoromethanesulfonate, samarium triflate, samarium iii triflate, samarium trifluoromethanesulfonate, trifluoromethanesulfonic acid samarium iii salt, samarium iii trifluoromethanesulfonate 97+%, sm otf 3, 3cf3o3s.sm, samarium 3+ tritriflate
PubChem CID 10145316
SMILES [Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 98%
Solubility Information Solubility in water: soluble
IUPAC Name samarium(3+);trifluoromethanesulfonate
Formula Weight 597.57
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Samarium(III) trifluoromethanesulfonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468793 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number383170100,383170010
Lot NumberA0468791 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number383170100,383170010
Lot NumberA0465567 Certificate TypeCertificate of Analysis Date27/08/2024 Catalog Number383170100,383170010
Lot NumberA0458198 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number383170100,383170010
Lot NumberA0395259 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number383170100,383170010
compliance-icons
Product Identifier
  • Samarium(III) trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Samarium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.