missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Samarium ingot/button,≈50.8mm (2.0 in.) dia., 99.5% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 Pc.
Description
Arylsulfonyl chlorides and arylsulfinates are reduced by a combination of samarium and titanium(IV) chloride to make diaryl disulfides. It is a neutron absorber, dopant for laser crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 7440-19-9 |
Molecular Formula | Sm |
MDL Number | MFCD00011233 MFCD00151299 |
Quantity | 1 Pc. |
Synonym | samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l |
Solubility Information | Insoluble in water. |
InChI Key | KZUNJOHGWZRPMI-UHFFFAOYSA-N |
SMILES | [Sm] |
IUPAC Name | samarium |
Molecular Weight (g/mol) | 150.36 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'