missing translation for 'onlineSavingsMsg'
Learn More
Samarium pieces, sublimed dendritic, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11361789

Samarium pieces, sublimed dendritic, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11361789
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11361789

Brand: Thermo Scientific Alfa Aesar 012423.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Color Gray
UN Number UN3178
Quantity 50 g
Percent Purity 99.9%
Odor Odorless
Form Pieces
Assay (REO)
Chemical Name or Material Samarium pieces, Sublimed dendritic

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB00X4975 Certificate TypeCertificate of Analysis Date22/03/2023 Catalog Number012423.09,012423.18
Lot NumberX27I023 Certificate TypeCertificate of Analysis Date15/11/2022 Catalog Number012375.A1,012375.C4,012375.A7,012423.18
Lot NumberW02I036 Certificate TypeCertificate of Analysis Date18/10/2022 Catalog Number012375.A1,012375.C4,012375.A7,012423.18
Lot NumberW02I035 Certificate TypeCertificate of Analysis Date18/10/2022 Catalog Number012375.A1,012375.C4,012375.A7,012423.18
Lot NumberR23I061 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog Number012375.A1,012375.C4,012375.A7,012423.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Samarium pieces, sublimed dendritic, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.