missing translation for 'onlineSavingsMsg'
Learn More

Samarium powder, -40 mesh, 99.9% (REO)

Product Code. 15451027
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 15451027

Brand: Thermo Scientific Alfa Aesar 012895.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-19-9 | Sm | 150.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Color Silver
Quantity 50 g
Mesh Size −40 Mesh
Odor Odorless
Packaging Ampule
Sensitivity Air and moisture sensitive
Form Powder
UN Number UN3089
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Samarium powder
Show More Show Less
compliance-icons
Product Identifier
  • Samarium powder
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
Hazard Statement
  • H228-Flammable solid.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Samarium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.