missing translation for 'onlineSavingsMsg'
Learn More

Samarium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Product Code. 11373848
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Quantity 50 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.96g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Samarium rod, 12.7mm (0.5 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'