Learn More
S Murray™ SAMCO™ Dissecting Scissors
Use these scissors for a variety of applications. S Murray™ SAMCO™ Dissecting Scissors feature fine points, open shanks and stainless steel construction.
Brand: S Murray™ E101/02
Description
- Stainless Steel dissecting scissors
Specifications
Dissecting Scissors | |
Straight | |
125 mm | |
Stainless Steel |
Product Content Correction
Product Title
S Murray™ SAMCO™ Dissecting Scissors > Blade Style: Straight; Length: 125mm
Spot an opportunity for improvement?Share a Content Correction