Learn More
S Murray™ SAMCO™ Dissecting Scissors
Use these scissors for a variety of applications. S Murray™ SAMCO™ Dissecting Scissors feature fine points, open shanks and stainless steel construction.
Brand:  S Murray™ E101/03
 View more versions of this product
Product Code. 12902055
  • £24.10 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Stainless Steel dissecting scissors
Specifications
Specifications
Dissecting Scissors
Straight
150 mm
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
S Murray™ SAMCO™ Dissecting Scissors > Blade Style: Straight; Length: 150mm