Learn More
Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes
Perform precise dispensing of volumes from 20 to 400 μL by using these exact volume transfer pipettes. Made of shatterproof, nontoxic polyethylene.
£26.60 - £380.00
Product Code | Brand | Description | Volume (Metric) Dispense | Length (Metric) | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) Dispense | Length (Metric) | Sterility | Price | Quantity & Availability | ||
15397803
|
Thermo Scientific™
191NL |
Samco Exact Volume Transfer Pipettes; 250μL Exact Volume Transfer Pipettes, Non-sterile (500/Pack, 5000/Case) | 250 μL | 10.19 cm | Non-sterile |
£44.05
Pack of 500 |
In Stock
Log in to see stock. |
||
15595243
|
Thermo Scientific™
783NL |
20μL Exact Volume Transfer Pipette | 20 μL | 5.3 cm | Non-sterile |
£45.95
Pack of 500 |
In Stock
Log in to see stock. |
||
15317903
|
Thermo Scientific™
784NL |
40μL Exact Volume Transfer Pipette | 40 μL | 5.9 cm | Non-sterile |
£43.75
Pack of 500 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
||
15327903
|
Thermo Scientific™
785NL |
60μL Exact Volume Transfer Pipette | 60 μL | 6.5 cm | Non-sterile |
£28.65
Pack of 500 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
||
15176284
|
Thermo Scientific™
786NL |
80μL Exact Volume Transfer Pipette | 80 μL | 7.5 cm | Non-sterile |
£279.00
Pack of 5000 |
In Stock
Log in to see stock. |
||
13450778
|
Thermo Scientific™
787NL |
100μL Exact Volume Transfer Pipette | 100 μL | 8.3 cm | Non-sterile |
£380.00
Pack of 5000 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
15337903
|
Thermo Scientific™
788NL |
150μL Exact Volume Transfer Pipette | 150 μL | 8.0 cm | Non-sterile |
£28.70
Pack of 500 |
In Stock
Log in to see stock. |
||
15397903
|
Thermo Scientific™
967NL |
400μL Exact Volume Transfer Pipette | 400 μL | 10.6 cm | Non-sterile |
£27.20
Pack of 500 |
In Stock
Log in to see stock. |
||
15367903
|
Thermo Scientific™
930NL |
50μL Exact Volume Transfer Pipette | 50 μL | 5.9 cm | Non-sterile |
£26.60
Pack of 500 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
15377903
|
Thermo Scientific™
941NL |
200μL Exact Volume Transfer Pipette | 200 μL | 8.89 cm | Non-sterile |
£38.80
Pack of 500 |
In Stock
Log in to see stock. |
||
15387903
|
Thermo Scientific™
966NL |
300μL Exact Volume Transfer Pipette | 300 μL | 9.5 cm | Non-sterile |
£37.15
Pack of 500 |
In Stock
Log in to see stock. |
||
Description
Perform precise dispensing of volumes from 20 to 400 μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
- Shatterproof, nontoxic polyethylene
- Nonsterile
- Low affinity surface
- Freezer ready
Specifications
Non-sterile |