Learn More
Thermo Scientific™ Samco™ Extra Long Transfer Pipettes
Perform safe and easy sampling from large containers using these extra-long transfer pipettes, perfect for research applications.
£36.80 - £235.19
Product Code | Brand | Description | Volume (Metric) | Length (Metric) | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Length (Metric) | Sterility | Price | Quantity & Availability | ||
15357853
|
Thermo Scientific™
262 |
Extra Long 9” Transfer Pipette | 5.6 mL | 22.5 cm | Non-sterile |
£38.70
Pack of 400 |
In Stock
Log in to see stock. |
||
15377853
|
Thermo Scientific™
262-1S |
Extra Long 9” Transfer Pipette | 5.6 mL | 22.5 cm | Sterile |
£60.80
Pack of 400 |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||
15387853
|
Thermo Scientific™
262-20S |
Extra Long 9” Transfer Pipette | 5.6 mL | 22.5 cm | Sterile |
£59.70
Pack of 400 |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||
15367853
|
Thermo Scientific™
262-10S |
Standard 9 in. Extra Long Transfer Pipet, 400 X 8 = 3200/CS | 6 mL | 22.5 cm | Sterile |
£42.50
Pack of 400 |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||
10111360
|
Thermo Scientific™
262NL |
Samco 6.0mL Extra Long Transfer Pipets, 9 in., Non-sterile, White Box | 6 mL | 22.5 cm | Non-sterile |
£38.44
Pack of 400 |
In Stock
Log in to see stock. |
||
11327873
|
Thermo Scientific™
262NL-10S |
Samco 6.0mL Extra Long Transfer Pipets, 9 in., Sterile, 10/pk, White Box | 6 mL | 22.5 cm | Sterile |
£68.60
Pack of 400 |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||
11317873
|
Thermo Scientific™
262NL-1S |
Samco 6.0mL Extra Long Transfer Pipets, 9 in., Sterile, Individually Packed, White Box | 6 mL | 22.5 cm | Sterile |
£60.80
Pack of 400 |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||
10705502
|
Thermo Scientific™
262NL-20S |
Samco 6.0mL Extra Long Transfer Pipets, 9 in., Sterile, 20/pk, White Box | 6 mL | 22.5 cm | Sterile |
£59.05
Pack of 400 |
In Stock
Log in to see stock. |
||
15242079
|
Thermo Scientific™
263 |
Extra Long 12” Transfer Pipette | 10.7 mL | 30.0 cm | Non-sterile |
£235.19
Pack of 1000 |
In Stock
Log in to see stock. |
||
15397853
|
Thermo Scientific™
263-1S |
Extra Long 12” Transfer Pipette | 10.7 mL | 30.0 cm | Sterile |
£77.60
Pack of 100 |
In Stock
Log in to see stock. |
||
15307863
|
Thermo Scientific™
263-20S |
Extra Long 12” Transfer Pipette | 10.7 mL | 30.0 cm | Sterile |
£68.70
Pack of 100 |
Estimated Shipment: 04-12-2024
Log in to see stock. |
||
11651839
|
Thermo Scientific™
263NL |
Samco 23.0mL Extra Long Transfer Pipets, 12 in., Non-sterile, White Box | 23 mL | 30 cm | Non-sterile |
£36.80
Pack of 100 |
In Stock
Log in to see stock. |
||
11337873
|
Thermo Scientific™
263NL-1S |
Samco 23.0mL Extra Long Transfer Pipets, 12 in., Sterile, Individually Packed, White Box | 23 mL | 30 cm | Sterile |
£75.55
Pack of 100 |
In Stock
Log in to see stock. |
||
11347873
|
Thermo Scientific™
263NL-20S |
Samco 23.0mL Extra Long Transfer Pipets, 12 in., Sterile, 20/pk, White Box | 23 mL | 30 cm | Sterile |
£69.30
Pack of 100 |
Estimated Shipment: 04-12-2024
Log in to see stock. |
||
Description
Perfect for research applications, these Extra-Long Transfer Pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, and autoclaving and prevents cross-contamination. They allow safe and easy sampling from large containers.
- Excellent for research applications
- General-purpose pipettes are ideal for routine sampling of liquids
- Well suited for use with large containers
Specifications
LDPE | |
No | |
Extra Long Transfer Pipet |