Learn More
Thermo Scientific™ Samco™ Transfer Pipettes
Prevent cross-contamination with these transfer pipettes, which have a built-in bulb that eliminates the need for plugging.
£15.30 - £388.00
Product Code | Brand | Volume (Metric) | Sterility | Bulb Draw (Metric) | Packaging Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Bulb Draw (Metric) | Packaging Type | Price | Quantity & Availability | |||
15307813
![]() |
Thermo Scientific™
202 |
7.7 mL | Non-sterile | 3.5 mL | Bulk 500/Pk |
£31.95
Pack of 500 |
In Stock
Log in to see stock. |
|||
15337813
![]() |
Thermo Scientific™
204-1S |
8.6 mL | Sterile | 6.0 mL | Individually Wrapped, 400/Pk |
£54.08
Pack of 400 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||
15307913
![]() |
Thermo Scientific™
B75-100 |
5.5 mL | Non-sterile | 2.4 mL | Bulk 500/Pk |
£21.29
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
11661839
![]() |
Thermo Scientific™
202NL |
7.7 mL | Non-sterile | 3.2 mL | Bulk, 500/pk |
£31.85
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
15778257
![]() |
Thermo Scientific™
202-10S |
7.7 mL | Sterile | 3.2 mL | 4000/Cs |
£388.00
Pack of 4000 |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||
11367873
![]() |
Thermo Scientific™
202NL-1S |
7.7 mL | Sterile | 3.2 mL | Individually Wrapped, 500/pk |
£93.25
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
12394508
![]() |
Thermo Scientific™
204NL |
8.0 mL | Non-sterile | 4.6 mL | Bulk 400/Pk |
£32.50
Pack of 400 |
In Stock
Log in to see stock. ![]() |
|||
15327883
![]() |
Thermo Scientific™
334NL |
3.1 mL | Non-sterile | 2.2 mL | Bulk 400/Pk |
£20.10
Pack of 400 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||
15317813
![]() |
Thermo Scientific™
202-1S |
7.7 mL | Sterile | 3.5 mL | Individually Wrapped 500/Pk |
£91.85
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
15327813
![]() |
Thermo Scientific™
202-20S |
7.7 mL | Sterile | 3.5 mL | 20/Bag, 25 Bags/Pk, 8 Pks/Cs. |
£39.15
Pack of 500 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||
10354470
![]() |
Thermo Scientific™
204 |
8.6 mL | Non-sterile | 6.0 mL | Bulk 400/Pk |
£34.20
Pack of 400 |
In Stock
Log in to see stock. |
|||
15357813
![]() |
Thermo Scientific™
204-20S |
8.6 mL | Sterile | 6.0 mL | 20/Bag, 400/Pk |
£39.15
Pack of 400 |
In Stock
Log in to see stock. ![]() |
|||
15252089
![]() |
Thermo Scientific™
273 |
1.8 mL | Non-sterile | 0.9 mL | Bulk 500/Pk |
£176.00
Pack of 4000 |
In Stock
Log in to see stock. |
|||
15337883
![]() |
Thermo Scientific™
335-11 |
7.5 mL | Non-sterile | 2.8 mL | Bulk 400/Pk |
£22.20
Pack of 400 |
In Stock
Log in to see stock. |
|||
15317863
![]() |
Thermo Scientific™
273-1S |
1.8 mL | Sterile | 0.9 mL | Individually Wrapped 500/Pk |
£53.25
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
15327863
![]() |
Thermo Scientific™
273-20S |
1.8 mL | Sterile | 0.9 mL | 20/Bag, 500/Pk |
£31.95
Pack of 500 |
Estimated Shipment: 05-09-2025
Log in to see stock. |
|||
15357883
![]() |
Thermo Scientific™
335-1S |
7.5 mL | Sterile | 2.8 mL | Individually Wrapped 400/Pk |
£67.05
Pack of 400 |
In Stock
Log in to see stock. ![]() |
|||
15367883
![]() |
Thermo Scientific™
335-20S |
7.5 mL | Sterile | 2.8 mL | 20/Bag, 500/Pk |
£45.95
Pack of 500 |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||
15397883
![]() |
Thermo Scientific™
336-1S |
4.9 mL | Sterile | 1.8 mL | Individually Wrapped 500/Pk |
£58.81
Pack of 500 |
In Stock
Log in to see stock. |
|||
15307893
![]() |
Thermo Scientific™
336-20S |
4.9 mL | Sterile | 1.8 mL | 20/Bag, 500/Pk |
£47.55
Pack of 500 |
Estimated Shipment: 15-08-2025
Log in to see stock. |
|||
15337893
![]() |
Thermo Scientific™
691 |
9.5 mL | Non-sterile | 6.2 mL | Bulk 250/Pk |
£15.30
Pack of 250 |
In Stock
Log in to see stock. |
|||
15347893
![]() |
Thermo Scientific™
691-1S |
9.5 mL | Sterile | 6.2 mL | Individually Wrapped 250/Pk |
£49.55
Pack of 250 |
In Stock
Log in to see stock. |
|||
15357893
![]() |
Thermo Scientific™
691-20S |
9.5 mL | Sterile | 6.2 mL | 20/Bag, 400/Pk |
£33.05
Pack of 400 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||
15307883
![]() |
Thermo Scientific™
325-11 |
1.05 mL | Non-sterile | 1.05 mL | Bulk 500/Pk |
£33.45
Pack of 500 |
In Stock
Log in to see stock. |
|||
15307873
![]() |
Thermo Scientific™
281NL |
0.6 mL | Non-sterile | 0.3 mL | Bulk, 500/Pk |
£22.05
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
15317883
![]() |
Thermo Scientific™
334-11 |
3.1 mL | Non-sterile | 2.2 mL | Bulk, 400/Pk. |
£20.10
Pack of 400 |
In Stock
Log in to see stock. |
|||
15377883
![]() |
Thermo Scientific™
336 |
4.9 mL | Nonsterile | 1.8 mL | Bulk 500/Pack |
£34.45
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
Description
Prevent cross-contamination with Thermo Scientific™ Samco™ Transfer Pipettes, which have a built-in bulb that eliminates the need for plugging, packaging and autoclaving. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
Recommended for: Research Applications
Specifications
LDPE |