missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ UniSampler Ex
Buerkle™ UniSampler Ex specially developed for sampling flammable liquids of Hazard Category AI (VbF, e.g. fuel).
Brand: Buerkle™ 5314-3000
This item is not returnable.
View return policy
Description
- Complete in a handy transport case incl. 4m hose (NBR), glass bottle 1000mL, bottle adapter, bottle stand, vacuum pump, earthing cable and hose-weight made of brass
- PTB approval D-333-009 618/00
- Glass bottle and hose are connected in such a way that they conduct electricity and are earthed via an earthing cable
- Sampler for liquid samples from precisely pre-determined depth, with hand-operated vacuum pump
- Sampling using vacuum pumps has the advantage that the liquid only comes into contact with the suction hose and the original sample bottle
Specifications
Manual Vacuum Sampler | |
NBR | |
Flammable Liquids of Hazard Category AI(VbF,e.g. Fuel) |
Borosilicate Glass, Polypropylene, PVC | |
1000 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buerkle™ UniSampler Ex > Capacity: 1000mL
Spot an opportunity for improvement?Share a Content Correction