Learn More
PureAire Monitoring Systems, Inc. Sample Draw Oxygen Deficiency Monitor with 10+ Year Sensor
Facilities using nitrogen, helium, and argon. The monitor is capable of sampling distances of up to 30 meters way and is commonly used for MRI’s, confined spaces, and cryogenics. Our durable zirconium sensor lasts 10 years and more.
Brand: PureAire Monitoring Systems, Inc.
Additional Details : Weight : 1.23010kg
Description
- Long-life 10+ year average sensor life
- Two-alarm setpoints for 19.5% and 18.0%
- Built-in audible alarm 90 db
- Plug-in power supply included
- Digital display, and 4-20mA analog output
- Wall mounting brackets
- Built-in flow sample pump
Why choose PureAire?
- As an industry leader in oxygen monitoring and gas detection, PureAire is dedicated to the safety of our clients, and the customers they serve, by manufacturing monitors that are long-lasting, innovative, and reliable.
What is an O2 Monitor?
- The Air Check O2 Deficiency Monitor is designed for continuous monitoring where low oxygen levels may pose a hazard to personnel.
What gases can an O2 monitor be used for?
- PureAire’s oxygen monitors can be used for nitrogen, argon, and helium gases.
Installation is easy. Plug-in ready.
- The monitor will arrive ready to work right out of the box. Simply secure the unit on the wall 3-5 feet from the gas source, and plug it in. The monitor will begin sampling oxygen levels after a 4 minute startup period. Levels are displayed clearly on the screen for easy access and visibility.
* Sampling tubes can be used in various locations including MRI Rooms
Certifications
CE Marked
Services
2 years
Specifications
CE | |
127 mm | |
5 x 7 x 5 in. | |
127 mm | |
-40°F to +104°F | |
2 Year after ETD | |
2 kg | |
178 mm |
5 in. | |
Oxygen | |
5 in. | |
0-100% RH | |
-40 to +40°C | |
4 lb. | |
7 in. |