Learn More
Spex CertiPrep™ Sample Prep Nesting Tray Set
Trays keep the samples secure during the grinding process
Brand: Spex CertiPrep™ 2189T
This item is not returnable.
View return policy
Description
A set of three nesting tray plates that allows titre plates to be securely held and stacked vertically on the 2189 adjustable clamp as used with the SPEX SamplePrep GenoGrinder 2010 homogeniser.- Nesting tray plates hold up to six deep-well titre plates securely vertically in the 2189 adjustable clamp
- Trays keep the samples secure during the grinding process
- Set comprises two nesting mid-trays and one bottom tray
Specifications
Nesting Tray Set | |
1 Pk. |
Product Content Correction
Product Title
Spex CertiPrep™ Sample Prep Nesting Tray Set > Product Type: Nesting Tray Set
Spot an opportunity for improvement?Share a Content Correction