Learn More
Spex CertiPrep™ Sample Prep Polycarbonate Cryovial
For use with 2199 Large Clamp Assembly for grinding media with SPEX SamplePrep homogeniser
Brand:  Spex CertiPrep™ 2252-PC-30
Product Code. 12859055
  • £131.00 / Pack of 30
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Polycarbonate cryovial with screw-on cap, 160mm x 121mm, 15mL. Holds grinding media up to 3/8in and fits SPEX SamplePrep homogeniser 2197 holder and 2661 cryo-block. For use with 2199 large clamp assembly. Grinding load per vial 6mL. Sold as a pack of 30 vials.
  • Holds grinding load of 6mL per vial
  • Polycarbonate so suitable for cryogenic grinding
Specifications
Specifications
15.8 mm
15 mL
30
101.6 mm
2199 Large Clamp Assembly
Polycarbonate
With Screw on Cap
Screw Cap
30 Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Spex CertiPrep™ Sample Prep Polycarbonate Cryovial > with cap, pre-cleaned in sturdy case, polycarbonate, can be filled and sealed individually 15mL SPEX SamplePrep