Learn More
Thermo Scientific™ Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™ sample and skimmer cones for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova series ICP-MS systems are available to suit your analysis requirements and designed for durability, performance and ease of use.
POA - £746.00
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
15457904
|
Thermo Scientific™
1311870 |
Skimmer Cone, Nickel, Insert-Version | iCAP Q/Qnova Series ICP-MS Systems |
£746.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17807066
|
Thermo Scientific™
1318480 |
Skimmer Cone Insert 3.5 (High Matrix) | iCAP Q/Qnova Series ICP-MS Systems |
£287.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16373348
|
Thermo Scientific™
BRE0006591 |
Skimmer Cone Insert 4.5 (Robust Plasma) | iCAP Q/Qnova Series ICP-MS Systems |
£181.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15261116
![]() |
Thermo Scientific™
1310900 |
Sample Cone Gasket | iCAP MX and iCAP Q/Qnova Series ICP-MS | N/A | ||||||
Description
Assure robust, uncomplicated trace elemental analysis for your laboratory with the Thermo Scientific iCAP MX and iCAP Q/Qnova series ICP-MS systems. Our easily changeable Ni and Pt sample and skimmer cones are available to suit your analysis requirements and designed for durability, performance and ease of use.
Select high-quality consumables for consistent performance. These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in either solid Ni construction or Pt tipped for even greater durability and specific analysis requirements.
- Sample cones made from nickel and platinum
- Skimmer cones made from nickel and platinum
- Platinum, boron free skimmer cone (for semiconductor-related applications)
- Skimmer cone inserts with different channel length to tailor instrument characteristics for each application