missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Sample Splitters

Product Code. 10432691 Shop All Retsch Products
Click to view available options
Depth (Metric):
6.3 mm
12.5 mm
25 mm
37.5 mm
50 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10432691

Brand: RETSCH 40.610.0002

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Product Type Sample Splitter
Depth (Metric) 12.5 mm
Includes 3 Collecting Receptacles 1.5L Each of Tin Plate, Stand of Coated Sheet Steel and Dividing Head Made of Stainless Steel
Particle Size 12.5 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.