Learn More
Buerkle™ Stainless Steel Sample Spoon
Buerkle™ Stainless Steel (AISI 304) Sample Spoon is a specially shaped spoon that allows you to reach almost every corner in a container and prevents samples from trickling out.
Brand: Buerkle™ 5320-0210
14.45 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Made of AISI 304 (1.4301) stainless steel
- Largely chemical resistant (not against acids and alkalis)
Specifications
Spoon | |
Autoclavable | |
210 mm |
Stainless Steel (V2A) | |
For Withdrawing Small Quantities of Samples, Foodstuffs |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.