Learn More
Thermo Scientific™ Sample Storage Starter Packs
GREENER_CHOICE
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs
Brand:  Thermo Scientific™ HHD-WP1D-2
 View more versions of this product
Product Code. 17886148
  • £2430.00 / 1 set
Explore more special offers
Description
Description
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs. Choose from five different starter pack options: BASIC, ADVANCED, PREMIUM, PREMIUM 0.5 mL and PREMIUM 1.mL. All Starter Packs include Matrix™ ScrewTop Tri-coded Tubes with an 8-Channel Screw Cap Decapper. Advanced and Premium Starter Packs also include a barcode reader.
Engineered with the value of your samples in mind:
  • Save 40% compared to buying the products individually.
  • Choose from five different starter pack options.
  • Eliminate the need to reformat non-2D cryotube samples.
  • Secure sample tracking with 2D, 1D, and human readable codes.
  • Cap/decap a full rack of tubes in less than 1 minute.
  • Scan 96 tubes in as little as 1 second with advanced and premium starter packs.
  • Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance.
Basic Starter Pack Includes:
    • 8-Channel Screw Cap Decapper (1 each)
    • 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
    • 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Advanced Starter Pack Includes:
    • 8-Channel Screw Cap Decapper (1 each)
    • VisionMate ST Single-Tube 2D Barcode Reader (1 each)
    • 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
    • 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Premium Starter Pack Includes:
    • 8-Channel Screw Cap Decapper (1 each)
    • VisionMate HSX High Speed Barcode Reader (1 each)
    • 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
    • 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Premium 0.5 mL Starter Pack Includes:
    • 8-Channel Screw Cap Decapper (1 each)
    • VisionMate HSX High Speed Barcode Reader (1 each)
    • 0.5 mL Matrix ScrewTop Tri-coded Tubes (5 cases)
Premium 1.0 mL Starter Pack Includes:
    • 8-Channel Screw Cap Decapper (1 each)
    • VisionMate HSX High Speed Barcode Reader (1 each)
    • 1.0 mL Matrix ScrewTop Tri-coded Tubes (5 cases)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tubes and Decapper
Basic Starter Pack