Learn More
Nickel Electro™ Aluminum Plate Sandbath Kit for use with HP1-3 Hotplates
Product Code. 11439499 Shop All Nickel-Electro Products
Click to view available options
Height (Metric):
27 mm
Unit Size:
Each
Product Code. 11439499
Brand: Nickel Electro™ SB13
Only null left
Add to basket
Add to basket
Nickel Electro™ Aluminum Plate Sandbath Kit converts flat top HP1-3 hotplates into sandbath for immersion of small samples.
  • Aluminum
  • Used to covert hotplates into sand baths
Specifications
Height (Metric) 27 mm