missing translation for 'onlineSavingsMsg'
Learn More
Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™ Product Code.: 10519770

Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™

Product Code. 10519770
3 kg
Click to view available options
Quantity:
1 kg
3 kg
This item is not returnable. View return policy

Product Code. 10519770

Brand: Fisher Chemical S/0360/63

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1610°C
Color Beige
Physical Form Solid
Quantity 3 kg
Formula Weight 60.08g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.