missing translation for 'onlineSavingsMsg'
Learn More

Sand, pure, 40-100 mesh, Thermo Scientific Chemicals

Product Code. 10267003
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Boiling Point 2230.0°C
Melting Point 1610.0°C
Color Beige to Gray
Physical Form Crystals
Quantity 250 g
Linear Formula SiO2
Merck Index 13,8567
Solubility Information Solubility in water: insoluble.
IUPAC Name dioxosilane
Formula Weight 60.08
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Sand, 40 to 100 Mesh
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'