missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, Pure, Silver, Fisher Chemical
Click to view available options
Quantity:
3 kg
Packaging:
Plastic powder jar
Specifications
Specifications
Boiling Point | 2230°C |
Melting Point | 1610°C |
CAS | 14808-60-7 |
Color | Beige |
Physical Form | Solid |
Quantity | 3 kg |
Molecular Formula | O2Si |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'