missing translation for 'onlineSavingsMsg'
Learn More

Sand, White Quartz, Honeywell Fluka™

Product Code. 15692910 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 lb.
Unit Size:
1kg
500g
50lb
5kg
This item is not returnable. View return policy

Product Code. 15692910

Brand: Honeywell-Fluka 2747395KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

50-70 mesh particle size

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color White
Quantity 5 kg
Linear Formula SiO2
UN Number NONH for all modes of transport
IUPAC Name dioxosilane
Formula Weight 60.08g/mol
Chemical Name or Material Sand, White Quartz
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.