missing translation for 'onlineSavingsMsg'
Learn More

Sand, White Quartz, Honeywell Fluka™

Product Code. 15607270
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a., powder

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2210°C
Melting Point 1710°C
Color White
Physical Form Solid
Quantity 5 kg
Linear Formula SiO2
UN Number NONH for all modes of transport
Formula Weight 60.08g/mol
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Sand, white quartz
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'